Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTACAGCAGGGGCAAGTGGTGGC[A/G]CTTATGGTAGCCAAATGATGGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300644 MIM: 300610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARMCX4 - armadillo repeat containing, X-linked 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GLA - galactosidase alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPH2 - heterogeneous nuclear ribonucleoprotein H2 (H') | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032393.2 | 1555 | Intron | NP_001027565.1 | |||
NM_019597.4 | 1555 | Intron | NP_062543.1 |
RPL36A-HNRNPH2 - RPL36A-HNRNPH2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199973.1 | 1555 | UTR 3 | NP_001186902.1 | |||
NM_001199974.1 | 1555 | UTR 3 | NP_001186903.1 |