Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGCAGACTGCGTGTGCAACGT[A/G]GAGTATTTCACTAGGGGAAAAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300846 MIM: 300171 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD99L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD99L2 - CD99 molecule like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184808.1 | 813 | Silent Mutation | TCC,TCT | S,S 171 | NP_001171737.1 | |
NM_001242614.1 | 813 | Silent Mutation | TCC,TCT | S,S 254 | NP_001229543.1 | |
NM_031462.3 | 813 | Silent Mutation | TCC,TCT | S,S 244 | NP_113650.2 | |
NM_134445.3 | 813 | Silent Mutation | TCC,TCT | S,S 172 | NP_604394.1 | |
NM_134446.3 | 813 | Silent Mutation | TCC,TCT | S,S 195 | NP_604395.1 | |
XM_011531202.2 | 813 | Intron | XP_011529504.1 | |||
XM_017029890.1 | 813 | Intron | XP_016885379.1 | |||
XM_017029891.1 | 813 | Intron | XP_016885380.1 |
MTMR1 - myotubularin related protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |