Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAGCTGCCATGGATGGGAGGCT[A/G]GGGTGAGATGGCCTGGAAGCAGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300189 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLG3 - discs large MAGUK scaffold protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166278.1 | 883 | Intron | NP_001159750.1 | |||
NM_020730.2 | 883 | Intron | NP_065781.1 | |||
NM_021120.3 | 883 | Silent Mutation | CTA,CTG | L,L 177 | NP_066943.2 | |
XM_005262248.3 | 883 | Intron | XP_005262305.1 | |||
XM_006724625.2 | 883 | Silent Mutation | CTA,CTG | L,L 177 | XP_006724688.1 | |
XM_006724626.2 | 883 | Silent Mutation | CTA,CTG | L,L 177 | XP_006724689.1 | |
XM_011530883.1 | 883 | Silent Mutation | CTA,CTG | L,L 177 | XP_011529185.1 | |
XM_017029322.1 | 883 | Intron | XP_016884811.1 | |||
XM_017029323.1 | 883 | Intron | XP_016884812.1 | |||
XM_017029324.1 | 883 | Intron | XP_016884813.1 | |||
XM_017029325.1 | 883 | Intron | XP_016884814.1 | |||
XM_017029326.1 | 883 | Intron | XP_016884815.1 | |||
XM_017029327.1 | 883 | Intron | XP_016884816.1 |
DLG3-AS1 - DLG3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |