Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCAGGGAAGATGGCTTCGTTTC[C/G]GAAGCTAACGCTTTCTGAAAAAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300616 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LUZP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LUZP4 - leucine zipper protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318840.1 | 57 | UTR 5 | NP_001305769.1 | |||
NM_016383.4 | 57 | Missense Mutation | CCG,CGG | P,R 5 | NP_057467.1 |