Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCTGAATGTAGACAGCCTTTTC[C/G]TAAAGTTACCCCCAGAAAAGAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300201 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYSLTR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYSLTR1 - cysteinyl leukotriene receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282186.1 | 1322 | Missense Mutation | ACG,AGG | T,R 303 | NP_001269115.1 | |
NM_001282187.1 | 1322 | Missense Mutation | ACG,AGG | T,R 303 | NP_001269116.1 | |
NM_001282188.1 | 1322 | Missense Mutation | ACG,AGG | T,R 303 | NP_001269117.1 | |
NM_006639.3 | 1322 | Missense Mutation | ACG,AGG | T,R 303 | NP_006630.1 |