Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTACACCATCCGAGAAGAAAT[G/T]CCAGAAAACGTCCTGATAGGCAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 400022 | ||||||||||||||||||||
Literature Links: |
PCDH11Y PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDH11Y - protocadherin 11 Y-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278619.1 | Intron | NP_001265548.1 | ||||
NM_032971.2 | Intron | NP_116753.1 | ||||
NM_032972.2 | Intron | NP_116754.1 | ||||
NM_032973.2 | Intron | NP_116755.1 | ||||
XM_017030079.1 | Intron | XP_016885568.1 | ||||
XM_017030080.1 | Intron | XP_016885569.1 | ||||
XM_017030081.1 | Intron | XP_016885570.1 | ||||
XM_017030082.1 | Intron | XP_016885571.1 | ||||
XM_017030083.1 | Intron | XP_016885572.1 |