Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCACTTGGCACTGGCACCTGGCC[A/G]AGGCACACTCTATGTCGGCGCAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300214 | ||||||||||||||||||||
Literature Links: |
PLXNB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLXNB3 - plexin B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163257.1 | 362 | Missense Mutation | CAA,CGA | Q,R 86 | NP_001156729.1 | |
NM_005393.2 | 362 | Missense Mutation | CAA,CGA | Q,R 63 | NP_005384.2 |