Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCATGGCCTTCATGTTGGGGGC[A/G]GCAATACGCTGAAGCACACTCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300040 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMC1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMC1A - structural maintenance of chromosomes 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281463.1 | 3337 | Silent Mutation | GCC,GCT | A,A 999 | NP_001268392.1 | |
NM_006306.3 | 3337 | Silent Mutation | GCC,GCT | A,A 1021 | NP_006297.2 |