Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTTTTCTTTTGTTTCTTTGGC[A/G]ATCTGTTCTATGTCTTTGTCATATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
USP51 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP51 - ubiquitin specific peptidase 51 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_201286.3 | 979 | Silent Mutation | ATC,ATT | I,I 300 | NP_958443.1 | |
XM_017029299.1 | 979 | Silent Mutation | ATC,ATT | I,I 300 | XP_016884788.1 | |
XM_017029300.1 | 979 | Silent Mutation | ATC,ATT | I,I 300 | XP_016884789.1 | |
XM_017029301.1 | 979 | Silent Mutation | ATC,ATT | I,I 300 | XP_016884790.1 |