Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTGAAGGAGCAGAGGCAGGCC[C/T]GGAAGGAGAGGCTCAGTGGGCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM199X PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM199X - family with sequence similarity 199, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207318.3 | 1248 | Missense Mutation | CGG,TGG | R,W 357 | NP_997201.1 | |
XM_005262079.2 | 1248 | Missense Mutation | CGG,TGG | R,W 314 | XP_005262136.1 | |
XM_017029270.1 | 1248 | Missense Mutation | CGG,TGG | R,W 174 | XP_016884759.1 |