Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGCCCTCTTCCACCAACAACAG[C/T]GGGGCATACAGGAGCCGACCCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300110 MIM: 313475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1F - calcium voltage-gated channel subunit alpha1 F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256789.2 | 2788 | Silent Mutation | CCA,CCG | P,P 1840 | NP_001243718.1 | |
NM_001256790.2 | 2788 | Silent Mutation | CCA,CCG | P,P 1786 | NP_001243719.1 | |
NM_005183.3 | 2788 | Silent Mutation | CCA,CCG | P,P 1851 | NP_005174.2 | |
XM_011543983.2 | 2788 | Silent Mutation | CCA,CCG | P,P 1779 | XP_011542285.1 | |
XM_017029836.1 | 2788 | Silent Mutation | CCA,CCG | P,P 901 | XP_016885325.1 |
SYP - synaptophysin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYP-AS1 - SYP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |