Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACCCAAATGCAGTCTACTCTGTC[C/T]GAGACCCCTCGCAGAGCTCAGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300685 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRRG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRRG3 - proline rich and Gla domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024082.3 | 260 | Nonsense Mutation | CGA,TGA | R,* 71 | NP_076987.3 | |
XM_005274747.3 | 260 | Nonsense Mutation | CGA,TGA | R,* 71 | XP_005274804.1 | |
XM_011531197.2 | 260 | Nonsense Mutation | CGA,TGA | R,* 71 | XP_011529499.1 |