Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAATCCTGTCAAAAAATGGAAG[G/T]AAAACCAGAAAATGAGAGTGAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEAL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCEAL9 - transcription elongation factor A like 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006612.1 | 248 | Missense Mutation | GGA,GTA | G,V 9 | NP_001006613.1 | |
NM_001006613.1 | 248 | Missense Mutation | GGA,GTA | G,V 9 | NP_001006614.1 | |
NM_001006614.1 | 248 | Missense Mutation | GGA,GTA | G,V 9 | NP_001006615.1 | |
NM_016303.2 | 248 | Missense Mutation | GGA,GTA | G,V 9 | NP_057387.1 |