Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTAGGTCCTGGCCCTGCTCCTGC[A/G]GCTTTGTAGACAGCCCCTGCATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300521 | ||||||||||||||||||||
Literature Links: |
GDPD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDPD2 - glycerophosphodiester phosphodiesterase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171191.1 | 200 | Missense Mutation | CAG,CGG | Q,R 29 | NP_001164662.1 | |
NM_001171192.1 | 200 | Missense Mutation | CAG,CGG | Q,R 108 | NP_001164663.1 | |
NM_001171193.1 | 200 | Missense Mutation | CAG,CGG | Q,R 29 | NP_001164664.1 | |
NM_017711.3 | 200 | Missense Mutation | CAG,CGG | Q,R 108 | NP_060181.2 | |
XM_011530977.1 | 200 | UTR 5 | XP_011529279.1 | |||
XM_017029614.1 | 200 | UTR 5 | XP_016885103.1 |
KIF4A - kinesin family member 4A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105373244 - uncharacterized LOC105373244 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |