Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCCCGCAGGCGCCACGCCAACC[A/G]CCGTGGGGACAGCTTCGGGGACAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
H2BFM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
H2BFM - H2B histone family member M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164416.1 | 376 | Missense Mutation | CAC,CGC | H,R 51 | NP_001157888.1 | |
XM_006724640.2 | 376 | Missense Mutation | CAC,CGC | H,R 51 | XP_006724703.1 | |
XM_011530922.2 | 376 | Missense Mutation | CAC,CGC | H,R 51 | XP_011529224.1 |