Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGAGAGAGAGCGAGACAGGGAA[C/T]GGGATCGGGACCGCGGGTATGACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300463 MIM: 314375 MIM: 300249 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PQBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PQBP1 - polyglutamine binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032381.1 | 554 | Missense Mutation | CGG,TGG | R,W 159 | NP_001027553.1 | |
NM_001032382.1 | 554 | Missense Mutation | CGG,TGG | R,W 159 | NP_001027554.1 | |
NM_001032383.1 | 554 | Missense Mutation | CGG,TGG | R,W 159 | NP_001027555.1 | |
NM_001032384.1 | 554 | Missense Mutation | CGG,TGG | R,W 159 | NP_001027556.1 | |
NM_001167989.1 | 554 | Missense Mutation | CGG,TGG | R,W 159 | NP_001161461.1 | |
NM_001167990.1 | 554 | Missense Mutation | CGG,TGG | R,W 151 | NP_001161462.1 | |
NM_001167992.1 | 554 | Intron | NP_001161464.1 | |||
NM_005710.2 | 554 | Missense Mutation | CGG,TGG | R,W 159 | NP_005701.1 | |
NM_144495.2 | 554 | Intron | NP_652766.1 | |||
XM_005272571.3 | 554 | Missense Mutation | CGG,TGG | R,W 159 | XP_005272628.1 | |
XM_005272572.4 | 554 | Intron | XP_005272629.1 | |||
XM_011543884.2 | 554 | Missense Mutation | CGG,TGG | R,W 159 | XP_011542186.1 | |
XM_017029207.1 | 554 | Missense Mutation | CGG,TGG | R,W 159 | XP_016884696.1 |
SLC35A2 - solute carrier family 35 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032289.2 | 554 | Intron | NP_001027460.1 | |||
NM_001042498.2 | 554 | Intron | NP_001035963.1 | |||
NM_001282647.1 | 554 | Intron | NP_001269576.1 | |||
NM_001282648.1 | 554 | Intron | NP_001269577.1 | |||
NM_001282649.1 | 554 | Intron | NP_001269578.1 | |||
NM_001282650.1 | 554 | Intron | NP_001269579.1 | |||
NM_001282651.1 | 554 | Intron | NP_001269580.1 | |||
NM_005660.2 | 554 | Intron | NP_005651.1 |
TIMM17B - translocase of inner mitochondrial membrane 17 homolog B (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |