Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGTACATAAGTCACGCTGGACAA[A/T]GAATGCTTTCTGAATGTAGACAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300201 | ||||||||||||||||||||
Literature Links: |
CYSLTR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYSLTR1 - cysteinyl leukotriene receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282186.1 | 1353 | Silent Mutation | TCA,TCT | S,S 313 | NP_001269115.1 | |
NM_001282187.1 | 1353 | Silent Mutation | TCA,TCT | S,S 313 | NP_001269116.1 | |
NM_001282188.1 | 1353 | Silent Mutation | TCA,TCT | S,S 313 | NP_001269117.1 | |
NM_006639.3 | 1353 | Silent Mutation | TCA,TCT | S,S 313 | NP_006630.1 |