Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCGGGAAATGAAGGATTATGGAC[C/T]AGCTAAGGGAGGCAAGAAGAAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300193 | ||||||||||||||||||||
Literature Links: |
HMGB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMGB3 - high mobility group box 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301228.1 | 346 | Missense Mutation | CCA,CTA | P,L 80 | NP_001288157.1 | |
NM_001301229.1 | 346 | Missense Mutation | CCA,CTA | P,L 80 | NP_001288158.1 | |
NM_001301231.1 | 346 | Missense Mutation | CCA,CTA | P,L 100 | NP_001288160.1 | |
NM_005342.3 | 346 | Missense Mutation | CCA,CTA | P,L 80 | NP_005333.2 |