Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTGGTGCGGCCACCCGGCGAGA[C/T]GGCTCCCAGGGCTTCACTGAAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300408 MIM: 300281 MIM: 300713 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIPAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIPAP1 - GRIP1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCND1 - potassium voltage-gated channel subfamily D member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004979.5 | 1828 | Missense Mutation | ATC,GTC | I,V 506 | NP_004970.3 | |
XM_011543910.2 | 1828 | Missense Mutation | ATC,GTC | I,V 506 | XP_011542212.1 | |
XM_017029508.1 | 1828 | Missense Mutation | ATC,GTC | I,V 417 | XP_016884997.1 | |
XM_017029509.1 | 1828 | Missense Mutation | ATC,GTC | I,V 417 | XP_016884998.1 | |
XM_017029510.1 | 1828 | Intron | XP_016884999.1 |
OTUD5 - OTU deubiquitinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |