Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCCCCACTCACTGTAGAAGGT[G/T]AGGTATCCAAAGGTTGCTGTGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300649 | ||||||||||||||||||||
Literature Links: |
SLC38A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC38A5 - solute carrier family 38 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033518.3 | 1120 | Silent Mutation | CTA,CTC | L,L 314 | NP_277053.2 | |
XM_005272694.3 | 1120 | Silent Mutation | CTA,CTC | L,L 361 | XP_005272751.2 | |
XM_005272695.4 | 1120 | Silent Mutation | CTA,CTC | L,L 361 | XP_005272752.3 | |
XM_005272697.2 | 1120 | Silent Mutation | CTA,CTC | L,L 320 | XP_005272754.2 | |
XM_005272698.4 | 1120 | Silent Mutation | CTA,CTC | L,L 314 | XP_005272755.2 | |
XM_006724569.3 | 1120 | Silent Mutation | CTA,CTC | L,L 314 | XP_006724632.1 | |
XM_017029960.1 | 1120 | Silent Mutation | CTA,CTC | L,L 320 | XP_016885449.1 | |
XM_017029961.1 | 1120 | Silent Mutation | CTA,CTC | L,L 314 | XP_016885450.1 |