Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGAGGCGCGCGCAGGAACGGGGC[A/G]GCCCAGCCTGTCTCCTTAAACACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300111 MIM: 313475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRICKLE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRICKLE3 - prickle planar cell polarity protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYP - synaptophysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003179.2 | 703 | Silent Mutation | GCC,GCT | A,A 229 | NP_003170.1 |
SYP-AS1 - SYP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |