Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTGTGTAGATTACTGAAGATTT[C/T]ATTGCTACATATCAGGCCAATTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300107 MIM: 300346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRS3 - bombesin receptor subtype 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HTATSF1 - HIV-1 Tat specific factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163280.1 | 623 | Silent Mutation | TTC,TTT | F,F 67 | NP_001156752.1 | |
NM_014500.4 | 623 | Silent Mutation | TTC,TTT | F,F 67 | NP_055315.2 | |
XM_005262404.3 | 623 | Silent Mutation | TTC,TTT | F,F 67 | XP_005262461.1 |