Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCGGTGCAGCATCTCCACCAAG[C/T]CCATGCTGCCCACCTGCTCGTGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300947 MIM: 300840 MIM: 300526 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC120 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC120 - coiled-coil domain containing 120 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRAF2 - PRA1 domain family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR45 - WD repeat domain 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029896.1 | 594 | Missense Mutation | NP_001025067.1 | |||
NM_007075.3 | 594 | Missense Mutation | NP_009006.2 |