Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTGGCTGTCACTGGCCCTGTAAT[C/T]GACAGCCCTGACTGCGTGCCCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLXNB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLXNB3 - plexin B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163257.1 | 453 | Silent Mutation | ATC,ATT | I,I 116 | NP_001156729.1 | |
NM_005393.2 | 453 | Silent Mutation | ATC,ATT | I,I 93 | NP_005384.2 |