Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGAAGAGACTTCTGAGCCTGAAT[C/G]TTCTTCTGAACAAACGACTGCAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC160 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC160 - coiled-coil domain containing 160 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101357.1 | 281 | Missense Mutation | TCT,TGT | S,C 31 | NP_001094827.1 | |
XM_005262409.4 | 281 | Missense Mutation | TCT,TGT | S,C 31 | XP_005262466.1 | |
XM_005262410.4 | 281 | Missense Mutation | TCT,TGT | S,C 31 | XP_005262467.1 |