Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCCTGATTTAGGCTTGCGTTC[C/T]TCTTCTACCTTTTTCTTTCCCAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300837 | ||||||||||||||||||||
Literature Links: |
DGKK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DGKK - diacylglycerol kinase kappa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013742.3 | 3778 | Silent Mutation | GAA,GAG | E,E 1231 | NP_001013764.1 | |
XM_017029268.1 | 3778 | Silent Mutation | GAA,GAG | E,E 1202 | XP_016884757.1 |