Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTTTGTTGCTGCCCATCTTTGA[G/T]TTTCAAGTGGTCTTCTGCTTGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP7D3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP7D3 - MAP7 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173516.1 | 2027 | Missense Mutation | ATC,CTC | I,L 634 | NP_001166987.1 | |
NM_001173517.1 | 2027 | Missense Mutation | ATC,CTC | I,L 617 | NP_001166988.1 | |
NM_024597.3 | 2027 | Missense Mutation | ATC,CTC | I,L 652 | NP_078873.2 | |
XM_005262472.1 | 2027 | Missense Mutation | ATC,CTC | I,L 651 | XP_005262529.1 | |
XM_017029843.1 | 2027 | Missense Mutation | ATC,CTC | I,L 638 | XP_016885332.1 |