Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCTAAAAGCTTTCCAACACAC[A/C]GGACATTCATAAGGTTTCTCTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 314993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF182 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF182 - zinc finger protein 182 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007088.1 | 1887 | Silent Mutation | CCG,CCT | P,P 526 | NP_001007089.1 | |
NM_001178099.1 | 1887 | Silent Mutation | CCG,CCT | P,P 545 | NP_001171570.1 | |
NM_006962.1 | 1887 | Silent Mutation | CCG,CCT | P,P 545 | NP_008893.1 |