Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTACTGAGGAGTTCAAATTCCT[C/T]AGGTGCCTTTTGAAGTACCTGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300777 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMLHE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMLHE - trimethyllysine hydroxylase, epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184797.1 | 1030 | Missense Mutation | AAG,GAG | K,E 287 | NP_001171726.1 | |
NM_018196.3 | 1030 | Missense Mutation | AAG,GAG | K,E 287 | NP_060666.1 | |
XM_011531182.2 | 1030 | Missense Mutation | AAG,GAG | K,E 236 | XP_011529484.1 | |
XM_017029620.1 | 1030 | Intron | XP_016885109.1 |