Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGAAATGAAGATTTGGAAGAGA[C/T]TGCTTCCACCGACCTGAAGTACCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300139 | ||||||||||||||||||||
Literature Links: |
IGBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGBP1 - immunoglobulin (CD79A) binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001551.2 | 510 | Missense Mutation | ACT,ATT | T,I 71 | NP_001542.1 | |
XM_017029489.1 | 510 | Missense Mutation | ACT,ATT | T,I 71 | XP_016884978.1 | |
XM_017029490.1 | 510 | Missense Mutation | ACT,ATT | T,I 71 | XP_016884979.1 |