Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGACCCCACAGAAACTCATAC[C/T]GTGCAGGATCACTGCCAGGCACCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300343 | ||||||||||||||||||||
Literature Links: |
MAGEA10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAGEA10 - MAGE family member A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011543.2 | 1311 | Missense Mutation | CAG,CGG | Q,R 293 | NP_001011543.2 | |
NM_001251828.1 | 1311 | Missense Mutation | CAG,CGG | Q,R 293 | NP_001238757.1 | |
NM_021048.4 | 1311 | Missense Mutation | CAG,CGG | Q,R 293 | NP_066386.2 |
MAGEA10-MAGEA5 - MAGEA10-MAGEA5 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204811.2 | 1311 | Intron | NP_001191740.1 |