Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCACCACCGCTCCATTGTGCCAC[A/G]TGTGTTCAAGGTGTGACTCAACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300050 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP11 - ubiquitin specific peptidase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004651.3 | 1139 | Missense Mutation | CAT,CGT | H,R 380 | NP_004642.2 | |
XM_005272674.3 | 1139 | Missense Mutation | CAT,CGT | H,R 107 | XP_005272731.1 | |
XM_011543988.1 | 1139 | Missense Mutation | CAT,CGT | H,R 107 | XP_011542290.1 |