Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGGACAGCAAACTTGATCTCATG[C/T]GGGGTCATCTACCAAAGGGACAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300798 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHKA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PHKA2 - phosphorylase kinase regulatory subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000292.2 | 3223 | Silent Mutation | CCA,CCG | P,P 1115 | NP_000283.1 | |
XM_005274548.4 | 3223 | Silent Mutation | CCA,CCG | P,P 1097 | XP_005274605.1 | |
XM_005274550.4 | 3223 | Silent Mutation | CCA,CCG | P,P 1087 | XP_005274607.1 | |
XM_006724496.3 | 3223 | Silent Mutation | CCA,CCG | P,P 1123 | XP_006724559.1 | |
XM_006724498.3 | 3223 | Silent Mutation | CCA,CCG | P,P 941 | XP_006724561.1 | |
XM_011545537.2 | 3223 | Silent Mutation | CCA,CCG | P,P 1090 | XP_011543839.1 | |
XM_011545538.2 | 3223 | Silent Mutation | CCA,CCG | P,P 784 | XP_011543840.1 | |
XM_017029580.1 | 3223 | Silent Mutation | CCA,CCG | P,P 821 | XP_016885069.1 |
PHKA2-AS1 - PHKA2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |