Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGTTTTTTATTTTTGGGGATGT[C/T]GGCAGGAGGAATTTCCTTTAAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300440 MIM: 312180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NKRF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NKRF - NFKB repressing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173487.1 | 1481 | Missense Mutation | AAC,GAC | N,D 243 | NP_001166958.1 | |
NM_001173488.1 | 1481 | Missense Mutation | AAC,GAC | N,D 228 | NP_001166959.1 | |
NM_017544.3 | 1481 | Missense Mutation | AAC,GAC | N,D 228 | NP_060014.2 | |
XM_011531365.2 | 1481 | Missense Mutation | AAC,GAC | N,D 228 | XP_011529667.1 |
UBE2A - ubiquitin conjugating enzyme E2 A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |