Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTGGGCCAGGAAGCCAAGAGCC[A/G]TTGCTGAAAAGATGGTGGTCAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 309860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAOB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAOB - monoamine oxidase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000898.4 | 1650 | Missense Mutation | ACG,ATG | T,M 505 | NP_000889.3 | |
XM_005272608.3 | 1650 | Missense Mutation | ACG,ATG | T,M 489 | XP_005272665.1 | |
XM_017029523.1 | 1650 | Missense Mutation | ACG,ATG | T,M 489 | XP_016885012.1 | |
XM_017029524.1 | 1650 | Missense Mutation | ACG,ATG | T,M 489 | XP_016885013.1 |