Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGCTGTCGGGAATGTGTTCCTC[A/G]TTGAACTTGACTGAGACTTCGTAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300017 | ||||||||||||||||||||
Literature Links: |
FLNA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLNA - filamin A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110556.1 | 6982 | Silent Mutation | AAC,AAT | N,N 2312 | NP_001104026.1 | |
NM_001456.3 | 6982 | Silent Mutation | AAC,AAT | N,N 2304 | NP_001447.2 | |
XM_011531127.1 | 6982 | Silent Mutation | AAC,AAT | N,N 2280 | XP_011529429.1 | |
XM_011531128.1 | 6982 | Silent Mutation | AAC,AAT | N,N 2272 | XP_011529430.1 | |
XM_011531129.1 | 6982 | Silent Mutation | AAC,AAT | N,N 2254 | XP_011529431.1 | |
XM_011531130.1 | 6982 | Silent Mutation | AAC,AAT | N,N 2246 | XP_011529432.1 | |
XM_011531131.1 | 6982 | Silent Mutation | AAC,AAT | N,N 2245 | XP_011529433.1 |
LOC105373386 - uncharacterized LOC105373386 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |