Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCTCAGTTTCCTTCCACCAGTG[A/G]TGGTTCTGGCTATAAGAATAACGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZCCHC12 - zinc finger CCHC-type containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312891.1 | 1470 | Missense Mutation | GAT,GGT | D,G 323 | NP_001299820.1 | |
NM_173798.3 | 1470 | Missense Mutation | GAT,GGT | D,G 323 | NP_776159.1 |