Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTTCGTAGTCTTCATCTCGAGG[C/T]TGTGTCCGAGGACTTTGTGTGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAB3 - TGF-beta activated kinase 1/MAP3K7 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152787.4 | 2604 | Silent Mutation | CAA,CAG | Q,Q 676 | NP_690000.3 | |
XM_005274483.3 | 2604 | Silent Mutation | CAA,CAG | Q,Q 648 | XP_005274540.2 | |
XM_005274484.2 | 2604 | Silent Mutation | CAA,CAG | Q,Q 642 | XP_005274541.2 | |
XM_011545488.2 | 2604 | Intron | XP_011543790.1 | |||
XM_017029399.1 | 2604 | Intron | XP_016884888.1 | |||
XM_017029400.1 | 2604 | Intron | XP_016884889.1 | |||
XM_017029401.1 | 2604 | Intron | XP_016884890.1 | |||
XM_017029402.1 | 2604 | Intron | XP_016884891.1 |