Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGATTGGGCGGGAATTACTAC[A/G]GATTGTTGTGTTGTTCCTGAAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300358 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WNK3 - WNK lysine deficient protein kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002838.3 | 5731 | Silent Mutation | TCC,TCT | S,S 1707 | NP_001002838.1 | |
NM_020922.4 | 5731 | Silent Mutation | TCC,TCT | S,S 1764 | NP_065973.2 | |
XM_006724593.3 | 5731 | Silent Mutation | TCC,TCT | S,S 1707 | XP_006724656.1 | |
XM_011530802.2 | 5731 | Silent Mutation | TCC,TCT | S,S 1717 | XP_011529104.1 | |
XM_017029741.1 | 5731 | Silent Mutation | TCC,TCT | S,S 1764 | XP_016885230.1 | |
XM_017029742.1 | 5731 | Silent Mutation | TCC,TCT | S,S 1764 | XP_016885231.1 | |
XM_017029743.1 | 5731 | Silent Mutation | TCC,TCT | S,S 1764 | XP_016885232.1 | |
XM_017029744.1 | 5731 | Silent Mutation | TCC,TCT | S,S 1764 | XP_016885233.1 | |
XM_017029745.1 | 5731 | Silent Mutation | TCC,TCT | S,S 1754 | XP_016885234.1 |