Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTCCATCTCAGCATGTCCCGCT[C/G]TCTCTGCACCTCCGCAAGGCTGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300313 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373310 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105373310 - NACHT, LRR and PYD domains-containing protein 10-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017030015.1 | 517 | Intron | XP_016885504.1 |
TEX13B - testis expressed 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031273.2 | 517 | Missense Mutation | CAG,GAG | Q,E 142 | NP_112563.1 |