Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGTGTCCGTGTACTTCACAAA[C/G]ACAGAATGGAAGCTTCTGGACCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZFP92 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFP92 - ZFP92 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136273.1 | 336 | Missense Mutation | AAC,AAG | N,K 25 | NP_001129745.1 | |
XM_005274652.3 | 336 | Missense Mutation | AAC,AAG | N,K 25 | XP_005274709.1 | |
XM_011531115.2 | 336 | Missense Mutation | AAC,AAG | N,K 25 | XP_011529417.1 | |
XM_017029281.1 | 336 | Missense Mutation | AAC,AAG | N,K 121 | XP_016884770.1 |