Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTTGATTAACCAGGCTATAAA[C/T]CAGGATGAAACCTTGGCCGTTTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RAP2C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAP2C - RAP2C, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271186.1 | 1165 | Missense Mutation | ATT,GTT | I,V 81 | NP_001258115.1 | |
NM_001271187.1 | 1165 | Missense Mutation | ATT,GTT | I,V 15 | NP_001258116.1 | |
NM_021183.4 | 1165 | Missense Mutation | ATT,GTT | I,V 81 | NP_067006.3 | |
XM_006724775.2 | 1165 | Missense Mutation | ATT,GTT | I,V 15 | XP_006724838.1 | |
XM_011531376.1 | 1165 | Missense Mutation | ATT,GTT | I,V 81 | XP_011529678.1 |
RAP2C-AS1 - RAP2C antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |