Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCACCCCGTGGCCCAGTGGCAT[C/T]TATTGCAACATCAATGATAGAATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300626 MIM: 311770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB11 - ankyrin repeat and SOCS box containing 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGA - phosphatidylinositol glycan anchor biosynthesis class A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002641.3 | 2306 | Intron | NP_002632.1 | |||
NM_020473.3 | 2306 | Intron | NP_065206.3 | |||
XM_011545539.2 | 2306 | Missense Mutation | AAT,GAT | N,D 225 | XP_011543841.1 | |
XM_017029581.1 | 2306 | Intron | XP_016885070.1 |