Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCCCAGTGTAGGCAGCATCAGG[A/G]CTGCAATGCACACCACTCCCAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300674 | ||||||||||||||||||||
Literature Links: |
MOSPD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MOSPD1 - motile sperm domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306188.1 | 972 | Intron | NP_001293117.1 | |||
NM_019556.2 | 972 | Missense Mutation | GCC,GTC | A,V 172 | NP_062456.1 | |
XM_011531366.2 | 972 | Missense Mutation | GCC,GTC | A,V 172 | XP_011529668.1 | |
XM_017029653.1 | 972 | Missense Mutation | GCC,GTC | A,V 172 | XP_016885142.1 | |
XM_017029654.1 | 972 | Intron | XP_016885143.1 |