Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGCTCTCCTTGCTTGCTGGTGGA[C/T]TGGCCACCTCTACATCCACTGCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300420 | ||||||||||||||||||||
Literature Links: |
PJA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PJA1 - praja ring finger ubiquitin ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032396.2 | 1512 | Missense Mutation | AAT,AGT | N,S 511 | NP_001027568.1 | |
NM_022368.4 | 1512 | Missense Mutation | AAT,AGT | N,S 378 | NP_071763.2 | |
NM_145119.3 | 1512 | Missense Mutation | AAT,AGT | N,S 566 | NP_660095.1 | |
XM_005262292.1 | 1512 | Missense Mutation | AAT,AGT | N,S 511 | XP_005262349.1 | |
XM_011531011.2 | 1512 | Missense Mutation | AAT,AGT | N,S 566 | XP_011529313.1 |