Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCAGCTCACGCCCCACTCCTCC[C/T]GCAGGCGCCGCCGCAGCCCTGCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602302 MIM: 615791 | ||||||||||||||||||||
Literature Links: |
HR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HR - hair growth associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005144.4 | 3961 | Missense Mutation | NP_005135.2 | |||
NM_018411.4 | 3961 | Intron | NP_060881.2 | |||
XM_005273569.1 | 3961 | Missense Mutation | XP_005273626.1 | |||
XM_006716367.1 | 3961 | Intron | XP_006716430.1 |
NUDT18 - nudix hydrolase 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |