Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCTTCATTTCGCTGCTGCTGTG[C/T]CTGTCGGGCATGTTCAGCGGCCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607803 | ||||||||||||||||||||
Literature Links: |
CNNM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNNM2 - cyclin and CBS domain divalent metal cation transport mediator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017649.4 | 989 | Silent Mutation | TGC,TGT | C,C 267 | NP_060119.3 | |
NM_199076.2 | 989 | Silent Mutation | TGC,TGT | C,C 267 | NP_951058.1 | |
NM_199077.2 | 989 | Silent Mutation | TGC,TGT | C,C 267 | NP_951059.1 | |
XM_005269933.4 | 989 | Silent Mutation | TGC,TGT | C,C 267 | XP_005269990.1 | |
XM_006717908.2 | 989 | Silent Mutation | TGC,TGT | C,C 267 | XP_006717971.1 | |
XM_011539911.2 | 989 | Silent Mutation | TGC,TGT | C,C 267 | XP_011538213.1 |
LOC107984265 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017017027.1 | 989 | Intron | XP_016872516.1 |