Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCACTGGGTCACCTCCAGGCCG[C/T]AGGAGCCTGCAGTCTTCTCGGCCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 195000 | ||||||||||||||||||||
Literature Links: |
ZP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZP1 - zona pellucida glycoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207341.3 | 285 | Nonsense Mutation | CAG,TAG | Q,* 89 | NP_997224.2 | |
XM_011544852.2 | 285 | Intron | XP_011543154.1 | |||
XM_011544853.2 | 285 | UTR 5 | XP_011543155.1 |