Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGTCAGAGCACTTTTTCCGATG[C/A]TGTTTGGATAAAAAATCACAAAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607585 | ||||||||||||||||||||
Literature Links: |
ATM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATM - ATM serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000051.3 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | NP_000042.3 | |
XM_005271561.4 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | XP_005271618.2 | |
XM_005271562.4 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | XP_005271619.2 | |
XM_006718843.3 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | XP_006718906.1 | |
XM_006718845.1 | 1863 | Nonsense Mutation | TGA,TGC | *,C 551 | XP_006718908.1 | |
XM_011542840.2 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | XP_011541142.1 | |
XM_011542842.2 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1844 | XP_011541144.1 | |
XM_011542843.2 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | XP_011541145.1 | |
XM_011542844.2 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1551 | XP_011541146.1 | |
XM_011542845.1 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1463 | XP_011541147.1 | |
XM_017017789.1 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | XP_016873278.1 | |
XM_017017790.1 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | XP_016873279.1 | |
XM_017017791.1 | 1863 | Nonsense Mutation | TGA,TGC | *,C 1899 | XP_016873280.1 | |
XM_017017792.1 | 1863 | Intron | XP_016873281.1 |
C11orf65 - chromosome 11 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152587.3 | 1863 | Intron | NP_689800.3 | |||
XM_005271412.3 | 1863 | Intron | XP_005271469.1 | |||
XM_005271413.3 | 1863 | Intron | XP_005271470.1 | |||
XM_005271414.3 | 1863 | Intron | XP_005271471.1 | |||
XM_005271415.4 | 1863 | Intron | XP_005271472.1 | |||
XM_011542639.2 | 1863 | Intron | XP_011540941.1 | |||
XM_011542640.2 | 1863 | Intron | XP_011540942.1 | |||
XM_011542641.2 | 1863 | Intron | XP_011540943.1 | |||
XM_011542642.1 | 1863 | Intron | XP_011540944.1 | |||
XM_011542643.2 | 1863 | Intron | XP_011540945.1 | |||
XM_017017246.1 | 1863 | Intron | XP_016872735.1 | |||
XM_017017247.1 | 1863 | Intron | XP_016872736.1 | |||
XM_017017248.1 | 1863 | Intron | XP_016872737.1 | |||
XM_017017249.1 | 1863 | UTR 3 | XP_016872738.1 | |||
XM_017017250.1 | 1863 | UTR 3 | XP_016872739.1 |