Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCTGTCCTTCCGGCTGAGCAGCT[T/C]CTCTGGCTTATCGAACTCAAGGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600509 MIM: 600937 | ||||||||||||||||||||
Literature Links: |
ABCC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCC8 - ATP binding cassette subfamily C member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000352.4 | 4819 | Missense Mutation | AAG,GAG | K,E 1564 | NP_000343.2 | |
NM_001287174.1 | 4819 | Missense Mutation | AAG,GAG | K,E 1565 | NP_001274103.1 | |
XM_011520331.2 | 4819 | Missense Mutation | AAG,GAG | K,E 1564 | XP_011518633.1 | |
XM_017018197.1 | 4819 | Missense Mutation | AAG,GAG | K,E 1587 | XP_016873686.1 | |
XM_017018198.1 | 4819 | Missense Mutation | AAG,GAG | K,E 1586 | XP_016873687.1 | |
XM_017018199.1 | 4819 | Missense Mutation | AAG,GAG | K,E 1586 | XP_016873688.1 | |
XM_017018200.1 | 4819 | Missense Mutation | AAG,GAG | K,E 1563 | XP_016873689.1 | |
XM_017018201.1 | 4819 | Intron | XP_016873690.1 | |||
XM_017018202.1 | 4819 | Missense Mutation | AAG,GAG | K,E 1086 | XP_016873691.1 | |
XM_017018203.1 | 4819 | Intron | XP_016873692.1 | |||
XM_017018204.1 | 4819 | Missense Mutation | AAG,GAG | K,E 883 | XP_016873693.1 |
KCNJ11 - potassium voltage-gated channel subfamily J member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |